CryoPlus™ Sample Storage System
Toimittaja: Thermo Fisher Scientific
7401 7403 7407
THBR7401EA 17120 EUR
THBR7401 THBR7403 BARN7407
CryoPlus™ Sample Storage System
Kryovarastointijärjestelmät
Increase valuable laboratory storage space with CryoPlus™ Storage Systems, which store up to 38500 vials (2,0 ml). The ideal combination of liquid nitrogen, storage reliability and microprocessor technology, the CryoPlus series LN₂ storage system provides precise and accurate control over all parameters without complicated programming.
  • 16 preset audible and visual alarm combinations provide maximum product protection
  • Automatic defogger incorporated into the manual fill index to improve visibility during retrieval
  • Liquid level sensor and fill port within