Pre-wetted cleanroom wipes, ThermaSat™ 60
Toimittaja: Texwipe
TX4304 STX4305 STX4306 TX4307 TX4308
TEXWTX4304EA 252 EUR
TEXWTX4304 TEXWSTX4305 TEXWSTX4306 TEXWTX4307 TEXWTX4308
Pre-wetted cleanroom wipes, ThermaSat™ 60
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Esikyllästetyt puhdastilan pyyhkeet
ThermaSat™ 60 wipes are made from 100% polyester with a sealed edge, and cleanroom laundered.
  • Wipe and clean surfaces, equipment, and parts
  • Apply and remove lubricants, adhesives, residues, and other solutions including disinfectants
  • Clean with solvents such as isopropyl alcohol (IPA), ethanol, acetone, and degreasers
  • Clean gloves, notebooks, phones, or any other item entering the cleanroom
  • Line trays for holding, protecting, drying, and storing of parts, equipment, and devices
  • Appropriate for use with temperatures less than 205 °C (400 °F) (dry wipes only)