Puhdastilan pyyhkeet, esikyllästetty, AlphaSat® with Vectra® AlphaSorb® 10
Toimittaja: Texwipe
TX1057
TEXWTX1057EA 1060 EUR
TEXWTX1057
Puhdastilan pyyhkeet, esikyllästetty, AlphaSat® with Vectra® AlphaSorb® 10
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Esikyllästetyt puhdastilan pyyhkeet
AlphaSat® with Vectra® AlphaSorb®10 incorporates ITW Texwipe's ultraclean AlphaSorb 10 sealed-border wipes (100% polyester) with 10% semiconductor-grade isopropyl alcohol (0,2 µm filtered) in an easy-to-use flatpack package.
  • Superior strength and abrasion resistance in a continuous-filament knitted substrate
  • Low particle generation and low extractable levels help prevent product and cleanroom contamination
  • Eliminates problems associated with purchasing, storing, filtering and diluting alcohol
  • Reduces alcohol consumption and eliminates over-spray of alcohol