Cleaning swabs, Foamtip™
Toimittaja: Texwipe
CF3050 CF1050 CF2050
TEXWCF3050EA 0 EUR
TEXWCF3050 TEXWCF1050 TEXWCF2050
Cleaning swabs, Foamtip™
Vanupuikot Näytteenottopuikot puhdistamiseen
These Foamtips™ swabs are constructed from medical grade polyurethane foam, which features the best open cell structure, providing particle entrapment characteristics.
  • Traps surface particles while cleaning
  • High solvent capacity and holds solvent well for high quantity applications
  • Foam thermally bonded to the swab handle without using adhesives
Used for cleaning excess adhesives after gluing, clean micro-mechanical devices and removes flux residues from printed circuit boards. Do not generate loose fibers or particles.
Varoitus: *Tällä sivullaMore Product Information