Probes for testo 440
Toimittaja: Testo
0615.5505 0615.1712 0635.1052 0635.1032 0635.1571 0635.1572 0635.9532 0635.9571 0635.9572 0635.9371 0635.9372 0635.9431 0635.9432 0636.9771 0636.9772 0636.9731 0636.9732 0636.9775 0635.0551 0632.1551 0632.1552 0632.1271 0632.1272 0628.0152 0635.1570 0632.1270 0635.9570 0635.9370 0635.9430 0636.9770 0636.9730 0632.1550 0615.1212
TEST0615.5505EA 56 EUR
TEST0615.5505 TEST0615.1712 620-2659 TEST0635.1032 620-2660 TEST0635.1572 TEST0635.9532 TEST0635.9571 TEST0635.9572 620-2661 TEST0635.9372 TEST0635.9431 TEST0635.9432 620-2662 TEST0636.9772 TEST0636.9731 TEST0636.9732 TEST0636.9775 TEST0635.0551 620-2658 TEST0632.1552 TEST0632.1271 TEST0632.1272 TEST0628.0152 TEST0635.1570 TEST0632.1270 TEST0635.9570 TEST0635.9370 TEST0635.9430 TEST0636.9770 TEST0636.9730 TEST0632.1550 TEST0615.1212
Probes for testo 440
Lämpöanturit
Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature. Humidity sensor is durable and traceable to international humidity standards, such as ILAC, PTB and NIST. Thus, even after years of use you can still count on measurement results that are reliable and correct.