Digital thermometer, testo 925
Toimittaja: Testo
0563.0925
TEST0563.0925EA 119 EUR
TEST0563.0925
Digital thermometer, testo 925
Lämpömittarit Digital Thermometers
This compact temperature measuring instrument testo 925 (TC Type K is the versatile solution for simple, fast and precise temperature measurements in industry and trade.
  • Easy, fast and precise temperature measurement with thermocouple Type K probe (1 TC Type K probe included)
  • Fast in-app configuration, graph history, second screen and measurement data memory in the testo Smart App
  • Wide range of applications due to large measuring range from ‒50 to 1000 °C
  • Large probe selection optional and compatible with commercially available TC Type K sensors
  • Audible