Clamp thermometer and heating kit operated with smartphone, Testo 115i
Toimittaja: Testo
0560.2115.02
TEST0560.2115.02EA 78 EUR
TEST0560.2115.02
Clamp thermometer and heating kit operated with smartphone, Testo 115i
Lämpömittarit Digital Thermometers
Compact professional measuring instrument from the Testo Smart Probes series for use with smartphones/tablets.
  • Measurement of flow and return temperature of heating systems
  • Temperature measurement on refrigeration systems for calculation of superheating and subcooling
  • Fast identification of temperature changes by graphic progression display
  • Measurement data analyzed and sent via testo Smart Probes App
  • Problem-free use at measuring points that are a long distance apart – Bluetooth® range up to 100 m
In conjunction with a smartphone or tablet, the handyMore Product Information