ISO Humidity Calibration Certificate
Toimittaja: Testo
0520.0166
TEST0520.0166EA 0 EUR
TEST0520.0166
ISO Humidity Calibration Certificate
Palvelut
This ISO calibration certificate certifies that your measuring instrument or probe is calibrated. Calibration is carried out according to the ISO standard.
  • Three calibration points: 11,3/50/75,3% RH at +25 °C
  • According to the ISO standard
  • Product/housing material: Paper
  • Product colour: White