Analysis and filtration paper, Tesorb
Toimittaja: TERVAKOSKI OY
2604120120 2604120260 2604135140 2604145160 2604165165 2604165165X 2604175175 2604175320 2604180180 2604185185 2604190190 2604200200 2604218341 2604219219 2604260320 2604315330 2604360700 430610250G 45045080G 25025080G 2604115315 2604385260 2604215910 270350250G 2604270420 3201020TK428
TERV2604120120EA 772 EUR
TERV2604120120 TERV2604120260 TERV2604135140 TERV2604145160 TERV2604165165 TERV2604165165X TERV2604175175 TERV2604175320 TERV2604180180 TERV2604185185 TERV2604190190 TERV2604200200 TERV2604218341 TERV2604219219 TERV2604260320 TERV2604315330 TERV2604360700 TERV430610250G TERV45045080G TERV25025080G TERV2604115315 TERV2604385260 TERV2604215910 TERV270350250G TERV2604270420 TERV3201020TK428
Analysis and filtration paper, Tesorb
Suodattimet Suodatinpaperit Kvalitatiivinen suodatinpaperi
An extremely pure paper perfectly manufactured for filtration, analysis and laboratory applications.
  • 80 – 300 g/m²
  • Made of 100% cotton linters and de-ionized process water only
  • Low ash content
  • No sizing, fillers or bindings
  • High content of α-cellulose
  • Consistent performance
100% de-ionized process water guarantees extremely pure quality.