Laskimokatetri, Versatus™
Toimittaja: TERUMO
SR-DU1445PX SR-DU1445WX SR-DU1645PX SR-DU1645WX SR-DU1745PX SR-DU1832PX SR-DU1832WX SR-DU1845PX SR-DU1845WX SR-DU2032PX SR-DU2032WX SR-DU2225PX SR-DU2225WX SR-DU2419PX SR-DU2619PX SR-DU2619WX SR-OF1464 SR-OF1651 SR-OF1832 SR-OF1851 SR-OF2032 SR-OF2051 SR-OF2225 SR-OF2419 SR-DU2419WD
TERUSR-DU1445PXEA 130 EUR
TERUSR-DU1445PX TERUSR-DU1445WX TERUSR-DU1645PX TERUSR-DU1645WX TERUSR-DU1745PX TERUSR-DU1832PX TERUSR-DU1832WX TERUSR-DU1845PX TERUSR-DU1845WX TERUSR-DU2032PX TERUSR-DU2032WX TERUSR-DU2225PX TERUSR-DU2225WX TERUSR-DU2419PX TERUSR-DU2619PX TERUSR-DU2619WX TERUSR-OF1464 TERUSR-OF1651 TERUSR-OF1832 TERUSR-OF1851 TERUSR-OF2032 TERUSR-OF2051 TERUSR-OF2225 TERUSR-OF2419 TERUSR-DU2419WD
Laskimokatetri, Versatus™
Neulat
Versatus cannulaes in three types: Straight, with wings, and with wings and injection port.
  • Catheter tip perfectly tapered to ensure a smooth transition from needle to catheter
  • PU is firm at room temperature but softens at body temperature
  • Ensuring a maximum of patient comfort
For almost each gauge, an obturator is available with the same colour which prevents thrombus formation in the lumen of the catheter when the infusion is discontinued.