Microplate racks for upright freezers
Toimittaja: TENAK
TE27522 TE27524 TE27532 TE27400 TE27402 TE27404 TE27406 TE27410 TE27412 TE27414 TE27416 TE27420 TE27422 TE27424 TE27426 TE27430 TE27432 TE27434 TE27440 TE27442 TE27450 TE27452 TE27454 TE27456 TE27458 TE27460 TE27462 TE27464 TE27466 TE27468 TE27470 TE27480 TE27482 TE27484 TE27486 TE27488 TE27490 TE27500 TE27502 TE27504 TE27506 TE27508 TE27510 TE27512 TE27514 TE27520 TE27526 TE27530 TE27534 TE27536 TE27540 TE27542 TE27544 TE27546 TE27548 TE27550 TE27552 TE27554 TE27556 TE27558 TE27560 TE27562 TE27564 TE27570 TE27572 TE27574 TE27576 TE27580 TE27582 TE27584 TE27586 TE27590 TE27592 TE27594 TE27596 TE27598 TE27600 TE27602 TE27604 TE27606 TE27610 TE27612 TE27614
TENATE27522EA 232.76 EUR
TENATE27522 479-1165 TENATE27532 TENATE27400 TENATE27402 TENATE27404 TENATE27406 TENATE27410 TENATE27412 TENATE27414 TENATE27416 TENATE27420 TENATE27422 TENATE27424 TENATE27426 TENATE27430 TENATE27432 TENATE27434 TENATE27440 TENATE27442 TENATE27450 TENATE27452 479-2183 TENATE27456 TENATE27458 TENATE27460 TENATE27462 479-2184 TENATE27466 TENATE27468 TENATE27470 TENATE27480 TENATE27482 TENATE27484 479-2185 TENATE27488 TENATE27490 TENATE27500 TENATE27502 479-2186 TENATE27506 TENATE27508 TENATE27510 479-2187 TENATE27514 TENATE27520 479-1166 TENATE27530 479-2188 TENATE27536 TENATE27540 TENATE27542 TENATE27544 479-2189 TENATE27548 TENATE27550 TENATE27552 479-1167 479-1168 TENATE27558 TENATE27560 479-2190 TENATE27564 TENATE27570 479-1163 TENATE27574 TENATE27576 TENATE27580 TENATE27582 TENATE27584 TENATE27586 TENATE27590 TENATE27592 TENATE27594 TENATE27596 TENATE27598 TENATE27600 479-1164 TENATE27604 TENATE27606 TENATE27610 TENATE27612 TENATE27614
Microplate racks for upright freezers
Koeputkitelineet Räkit kuoppalevyille
Complete range of racks for microtiter plates for use in upright freezers, made of stainless steel. The racks can either be used as one plate per shelf or the plates can be stacked.