Racks for chest freezers, Eco AluRack
Toimittaja: TENAK
TE11566 TE11514 TE11518 TE11458 TE11458B TE11460 TE11460B TE11462 TE11462B TE11464 TE11464B TE11465 TE11465B TE11466 TE11466B TE11468 TE11468B TE11470 TE11470B TE11500 TE11500B TE11502 TE11502B TE11504 TE11504B TE11506 TE11506B TE11508 TE11508B TE11510 TE11510B TE11512 TE11512B TE11514B TE11516 TE11516B TE11518B TE11520 TE11520B TE11522 TE11522B TE11530 TE11530B TE11532 TE11532B TE11534 TE11534B TE11536 TE11536B TE11538 TE11538B TE11540 TE11540B TE11542 TE11542B TE11544 TE11544B TE11550 TE11552 TE11560 TE11560B TE11562 TE11562B TE11564 TE11564B TE11566B TE11570 TE11570B TE11572 TE11572B TE11574 TE11574B TE11576 TE11576B
TENATE11566EA 73.69 EUR
TENATE11566 TENATE11514 TENATE11518 TENATE11458 TENATE11458B TENATE11460 TENATE11460B TENATE11462 TENATE11462B TENATE11464 TENATE11464B TENATE11465 TENATE11465B TENATE11466 TENATE11466B TENATE11468 TENATE11468B TENATE11470 TENATE11470B TENATE11500 TENATE11500B TENATE11502 TENATE11502B TENATE11504 TENATE11504B TENATE11506 TENATE11506B TENATE11508 TENATE11508B TENATE11510 TENATE11510B TENATE11512 TENATE11512B TENATE11514B TENATE11516 TENATE11516B TENATE11518B TENATE11520 TENATE11520B TENATE11522 TENATE11522B TENATE11530 TENATE11530B TENATE11532 TENATE11532B TENATE11534 TENATE11534B TENATE11536 TENATE11536B TENATE11538 TENATE11538B TENATE11540 TENATE11540B TENATE11542 TENATE11542B TENATE11544 TENATE11544B TENATE11550 TENATE11552 TENATE11560 TENATE11560B TENATE11562 TENATE11562B TENATE11564 TENATE11564B TENATE11566B TENATE11570 TENATE11570B TENATE11572 TENATE11572B TENATE11574 TENATE11574B TENATE11576 TENATE11576B
Racks for chest freezers, Eco AluRack
Koeputkitelineet Telineet kryolaatikoille
The rack is made of seawater resistant aluminum and fits into any freezer and can be used with all standard cryoboxes.
These racks are available in six different heights with and without cryoboxes.