Racks for upright freezers, Eco AluRack
Toimittaja: TENAK
TE11252 TE11232 TE11238 TE11278 TE11348 TE11344 TE11254 TE11254B TE11256 TE11256B TE11260 TE11262 TE11262B TE11264 TE11264B TE11266 TE11266B TE11272 TE11272B TE11274 TE11274B TE11276 TE11276B TE11278B TE11284 TE11284B TE11286 TE11286B TE11288 TE11288B TE11294 TE11294B TE11296 TE11296B TE11302 TE11302B TE11304 TE11304B TE11310 TE11310B TE11312 TE11312B TE11320 TE11322 TE11330 TE11332 TE11334 TE11340 TE11342 TE11342B TE11344B TE11346 TE11346B TE11348B TE11350 TE11350B TE11352 TE11352B TE11120 TE11120B TE11122 TE11122B TE11124 TE11124B TE11126 TE11126B TE11130 TE11130B TE11132 TE11132B TE11134 TE11134B TE11136 TE11136B TE11138 TE11138B TE11140 TE11140B TE11150 TE11150B TE11152 TE11152B TE11154 TE11154B TE11156 TE11156B TE11220 TE11220B TE11222 TE11222B TE11224 TE11224B TE11230 TE11230B TE11232B TE11234 TE11234B TE11250 TE11236 TE11236B TE11238B TE11250B TE11252B
479-1322EA 100.59 EUR
479-1322 TENATE11232 TENATE11238 TENATE11278 TENATE11348 TENATE11344 TENATE11254 TENATE11254B TENATE11256 TENATE11256B TENATE11260 TENATE11262 TENATE11262B TENATE11264 TENATE11264B TENATE11266 TENATE11266B TENATE11272 TENATE11272B TENATE11274 TENATE11274B TENATE11276 TENATE11276B TENATE11278B TENATE11284 TENATE11284B TENATE11286 TENATE11286B TENATE11288 TENATE11288B TENATE11294 TENATE11294B TENATE11296 TENATE11296B TENATE11302 TENATE11302B TENATE11304 TENATE11304B TENATE11310 TENATE11310B TENATE11312 TENATE11312B TENATE11320 TENATE11322 TENATE11330 TENATE11332 TENATE11334 TENATE11340 TENATE11342 TENATE11342B TENATE11344B TENATE11346 TENATE11346B TENATE11348B TENATE11350 TENATE11350B TENATE11352 TENATE11352B TENATE11120 TENATE11120B TENATE11122 TENATE11122B TENATE11124 TENATE11124B TENATE11126 TENATE11126B TENATE11130 TENATE11130B TENATE11132 TENATE11132B TENATE11134 TENATE11134B TENATE11136 TENATE11136B TENATE11138 TENATE11138B TENATE11140 TENATE11140B TENATE11150 TENATE11150B TENATE11152 TENATE11152B TENATE11154 TENATE11154B TENATE11156 TENATE11156B TENATE11220 TENATE11220B TENATE11222 TENATE11222B TENATE11224 TENATE11224B TENATE11230 TENATE11230B TENATE11232B TENATE11234 TENATE11234B TENATE11250 TENATE11236 TENATE11236B TENATE11238B TENATE11250B TENATE11252B
Racks for upright freezers, Eco AluRack
Koeputkitelineet Telineet kryolaatikoille
These Eco AluRack fits into any freezer and can be used with all standard cryoboxes and assembled without netting or welding.
  • Low weight aluminium
  • Side opening design
  • Available with or without cryoboxes