Add-on storage units
Toimittaja: TEKNOMEK
TCA0100 TCA0200 TCA0300 TCA0600 TCA0700
TEKMTCA0100EA 0 EUR
TEKMTCA0100 TEKMTCA0200 TEKMTCA0300 TEKMTCA0600 TEKMTCA0700
Add-on storage units
Kalusteet Säilytyskaapit
304 grade stainless steel add on unit for rear railed workbench is perfect for adding additional storage. Cannot be sold singly.
  • Fully welded 304-grade stainless steel
  • Brushed dull polish finish
Supplied fitted to workbench when ordered together. Choice of one drawer, two drawers, three drawers or cupboard unit, for our rear railed workbenches. Enables the user to create a workstation to suit their individual requirements.