Zirconium(IV) acetylacetonate

Toimittaja: TCI
Varoitus

Synonyms: Tetrakis(2,4-pentanedionato)zirconium(IV), Zirconium(IV) 2,4-pentanedionate

Z0006-25G Z0006-500G Z0006-100G
TCIAZ0006-25GEA 36.3 EUR
TCIAZ0006-25G TCIAZ0006-500G TCIAZ0006-100G
Zirconium(IV) acetylacetonate
Zirconium(IV) acetylacetonate
Kaava: C₂₀H₂₈O₈Zr
MDL numero: MFCD00000036
CAS-numero: 17501-44-9
EINECS: 241-510-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60252cf96431e58f99f9a2abf84d237b01bcc331&vid=60252cf96431e58f99f9a2abf84d237b01bcc331&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Melting point (Decomposition) 188.0 to 195.0 deg-C
Solubility in hot Toluene very faint turbidity
Ignition residue 24.0 to 26.2 % (as ZrO2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR