3',4',5'-Trifluoroacetophenone ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

T3825-5G T3825-25G
TCIAT3825-5GEA 47.3 EUR
TCIAT3825-5G TCIAT3825-25G
3',4',5'-Trifluoroacetophenone ≥98.0% (by GC)
3',4',5'-Trifluoroacetophenone
Kaava: C₈H₅F₃O
MDL numero: MFCD00083533
CAS-numero: 220141-73-1

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR