Tina(II)bromidi ≥97.0% (by titrimetric analysis) for perovskite precursor

Toimittaja: TCI

T3573-1G T3573-5G
TCIAT3573-1GEA 33 EUR
TCIAT3573-1G TCIAT3573-5G
Tina(II)bromidi ≥97.0% (by titrimetric analysis) for perovskite precursor
Tina(II)bromidi
Kaava: Br₂Sn
MDL numero: MFCD00011239
CAS-numero: 10031-24-0

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (Iodometric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR