1H,1H,2H,2H-Perfluorooctyltrimethoxysilane ≥97.0% (by GC)

Toimittaja: TCI
Varoitus

T3560-5G T3560-25G
TCIAT3560-5GEA 90.2 EUR
TCIAT3560-5G TCIAT3560-25G
1H,1H,2H,2H-Perfluorooctyltrimethoxysilane ≥97.0% (by GC)
1H,1H,2H,2H-Perfluorooctyltrimethoxysilane
Kaava: C₁₁H₁₃F₁₃O₃Si
Kiehumispiste: 71°C
MDL numero: MFCD00153261
CAS-numero: 85857-16-5
EINECS: 288-657-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57146b72527b2bf2e4d438cf10dc4af695c9c97b&vid=57146b72527b2bf2e4d438cf10dc4af695c9c97b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR