2-[4-(Trifluoromethyl)phenyl]ethylamine ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Vaara

T3278-1G T3278-5G
TCIAT3278-1GEA 33 EUR
TCIAT3278-1G TCIAT3278-5G
2-[4-(Trifluoromethyl)phenyl]ethylamine ≥98.0% (by GC, titration analysis)
2-[4-(Trifluoromethyl)phenyl]ethylamine
Kaava: C₉H₁₀F₃N
Kiehumispiste: 90°C
Storage Temperature: Jääkaappi
MDL numero: MFCD01529875
CAS-numero: 775-00-8
UN: 2735
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR