1-(p-Tolyl)-1-cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Toimittaja: TCI

T3097-5G T3097-25G
TCIAT3097-5GEA 77 EUR
TCIAT3097-5G TCIAT3097-25G
1-(p-Tolyl)-1-cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-(p-Tolyl)-1-cyclohexanecarboxylic acid
Kaava: C₁₄H₁₈O₂
MDL numero: MFCD00065264
CAS-numero: 84682-27-9
EINECS: 283-589-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dfcd91f3940af808e9c8d37fefcba04954dadaf&vid=6dfcd91f3940af808e9c8d37fefcba04954dadaf&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 169.0 to 173.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR