3-(2-Thienyl)pyridine ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

T3055-200MG T3055-1G
TCIAT3055-200MGEA 61.6 EUR
TCIAT3055-200MG TCIAT3055-1G
3-(2-Thienyl)pyridine ≥98.0% (by GC)
3-(2-Thienyl)pyridine
Kaava: C₉H₇NS
Kiehumispiste: 100°C
MDL numero: MFCD01860052
CAS-numero: 21298-53-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74fd2c800231ad57328c6f4eabf8fc2564297aee&vid=74fd2c800231ad57328c6f4eabf8fc2564297aee&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR