tri-Magnesium dicitrate nonahydrate ≥98.0% (by titrimetric analysis)

Toimittaja: TCI

T2966-25G T2966-500G
TCIAT2966-25GEA 17.6 EUR
TCIAT2966-25G TCIAT2966-500G
tri-Magnesium dicitrate nonahydrate ≥98.0% (by titrimetric analysis)
tri-Magnesium dicitrate nonahydrate
Kaava: C₁₂H₁₀Mg₃O₁₄·9H₂O
MDL numero: MFCD00036422
CAS-numero: 153531-96-5
EINECS: 222-093-9
Merck Index: 14,05663
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dec3ae5ba35cace20e5b12fe1c7934b7975195d5&vid=dec3ae5ba35cace20e5b12fe1c7934b7975195d5&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Water 25.0 to 28.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR