Tin(IV) 2,3-naphthalocyanine dichloride

Toimittaja: TCI
Vaara

T2940-200MG T2940-1G
TCIAT2940-200MGEA 114 EUR
TCIAT2940-200MG TCIAT2940-1G
Tin(IV) 2,3-naphthalocyanine dichloride
Tin(IV) 2,3-naphthalocyanine dichloride
Kaava: C₄₈H₂₄Cl₂N₈Sn
MDL numero: MFCD00171716
CAS-numero: 26857-61-4
UN: 3146
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e0b88cbef86cd54f181fed6ff94541b4a9f2f17&vid=8e0b88cbef86cd54f181fed6ff94541b4a9f2f17&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Green to Dark green powder to crystal
Elemental analysis (Carbon) 60.70 to 67.10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR