Tilorone dihydrochloride ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

T2836-1G T2836-100MG
TCIAT2836-1GEA 222 EUR
TCIAT2836-1G TCIAT2836-100MG
Tilorone dihydrochloride ≥98.0% (by HPLC, titration analysis)
Tilorone dihydrochloride
Kaava: C₂₅H₃₄N₂O₃
Sulamispiste: 234°C
MDL numero: MFCD00134071
CAS-numero: 27591-69-1
Merck Index: 14,09440

Order Now

Testitulosten erittely

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR