Thiamphenicol ≥98.0% (by HPLC, total nitrogen)

Toimittaja: TCI

T2802-5G T2802-25G
TCIAT2802-5GEA 57.2 EUR
TCIAT2802-5G TCIAT2802-25G
Thiamphenicol ≥98.0% (by HPLC, total nitrogen)
Thiamphenicol
Kaava: C₁₂H₁₅Cl₂NO₅S
MDL numero: MFCD00467983
CAS-numero: 15318-45-3
EINECS: 239-355-3
Merck Index: 14,09301

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 164.0 to 168.0 deg-C
Specific rotation [a]20/D +10.0 to +13.0 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR