4-(Trimethylsilyl)phenylboronic acid (contains varying amounts of Anhydride)

Toimittaja: TCI
Varoitus

T2664-1G T2664-5G
TCIAT2664-1GEA 47.3 EUR
TCIAT2664-1G TCIAT2664-5G
4-(Trimethylsilyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(Trimethylsilyl)phenylboronic acid
Kaava: C₉H₁₅BO₂Si
MDL numero: MFCD00093348
CAS-numero: 17865-11-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=655fb4d2f0fa825880c6f79ccdbde89f204211f6&vid=655fb4d2f0fa825880c6f79ccdbde89f204211f6&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR