Trimethylsilylmethylmagnesium chloride ∼18% ∼1 mol/L in THF

Toimittaja: TCI
Vaara

T2609-100ML
TCIAT2609-100MLEA 114 EUR
TCIAT2609-100ML
Trimethylsilylmethylmagnesium chloride ∼18% ∼1 mol/L in THF
Trimethylsilylmethylmagnesium chloride
Kaava: C₄H₁₁ClMgSi
MDL numero: MFCD00009916
CAS-numero: 13170-43-9
UN: 3399
ADR: 4.3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=173d65769a8d316997b69bd20c19f718c3e054cf&vid=eed1265331152425d6283ddf6abfd0e19a4e72db&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Concentration (Neutralization titration) 18.5 to 20.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR