EDTA trinatriumsuola H2O ≥98.0% (by titrimetric analysis) for biochemical research

Toimittaja: TCI

T2599-25G T2599-5G
TCIAT2599-25GEA 37.4 EUR
TCIAT2599-25G TCIAT2599-5G
EDTA trinatriumsuola H2O ≥98.0% (by titrimetric analysis) for biochemical research
EDTA trinatriumsuola H2O
Kaava: C₁₀H₁₃N₂Na₃O₈·H₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013305
CAS-numero: 85715-60-2
EINECS: 205-758-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d9b57e5f7a199fa3be86f25667d9e5fc3e6c976&vid=6b571b19de4b0c95e436c21fde5ff445d4e55d1c&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (as Dihydrate)
Water 7.5 to 10.5 %
Enzyme inhibition test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR