Cyclopentyltrichlorosilane ≥95.0% (by GC)

Toimittaja: TCI
Vaara

T2576-5G T2576-25G
TCIAT2576-5GEA 27.5 EUR
TCIAT2576-5G TCIAT2576-25G
Cyclopentyltrichlorosilane ≥95.0% (by GC)
Cyclopentyltrichlorosilane
Kaava: C₅H₉Cl₃Si
Kiehumispiste: 178°C
Tiheys: 1,2756 g/cm³ (20 °C)
Leimahduspiste: 68°C
MDL numero: MFCD00236508
CAS-numero: 14579-03-4
EINECS: 238-621-6
UN: 2987
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14b45deefbd2c4278556737968a581212de5ceb5&vid=14b45deefbd2c4278556737968a581212de5ceb5&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR