Tamoxifen citrate ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Vaara

T2510-1G T2510-5G
TCIAT2510-1GEA 91.3 EUR
TCIAT2510-1G TCIAT2510-5G
Tamoxifen citrate ≥98.0% (by HPLC, titration analysis)
Tamoxifen citrate
Kaava: C₂₆H₂₉NO
Sulamispiste: 142°C
Storage Temperature: Jääkaappi
MDL numero: MFCD00058321
CAS-numero: 54965-24-1
EINECS: 259-415-2
Merck Index: 14,09048

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR