trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

T2424-1G T2424-5G
TCIAT2424-1GEA 35.2 EUR
TCIAT2424-1G TCIAT2424-5G
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid
Kaava: C₈H₁₁F₃O₂
MDL numero: MFCD09037363
CAS-numero: 133261-33-3

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR