Tri-p-tolylbismuthine ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI

T2261-1G
TCIAT2261-1GEA 147 EUR
TCIAT2261-1G
Tri-p-tolylbismuthine ≥98.0% (by HPLC, titration analysis)
Tri-p-tolylbismuthine
Kaava: C₂₁H₂₁Bi
CAS-numero: 5142-75-6

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Chelometric Titration) min. 98.0 %
Melting point 117.0 to 121.0 deg-C
Solubility in THF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR