1-(Trifluoroacetyl)piperidine ≥98.0% (by GC)

Toimittaja: TCI

T2087-25G T2087-5G
TCIAT2087-25GEA 195 EUR
TCIAT2087-25G TCIAT2087-5G
1-(Trifluoroacetyl)piperidine ≥98.0% (by GC)
1-(Trifluoroacetyl)piperidine
Kaava: C₇H₁₀F₃NO
Kiehumispiste: 77°C
Leimahduspiste: 76°C
MDL numero: MFCD00466213
CAS-numero: 340-07-8

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR