(3-Isocyanatopropyl)trimethoxysilane ≥97.0% (by GC)

Toimittaja: TCI
Vaara

T2044-100G T2044-25G
TCIAT2044-100GEA 108 EUR
TCIAT2044-100G TCIAT2044-25G
(3-Isocyanatopropyl)trimethoxysilane ≥97.0% (by GC)
(3-Isocyanatopropyl)trimethoxysilane
Kaava: C₇H₁₅NO₄Si
Kiehumispiste: 75°C
Leimahduspiste: 108°C
MDL numero: MFCD00015793
CAS-numero: 15396-00-6
EINECS: 239-415-9
UN: 2927
ADR: 6.1,II

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR