Tributylsilane ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

T2043-25G T2043-5G
TCIAT2043-25GEA 286 EUR
TCIAT2043-25G TCIAT2043-5G
Tributylsilane ≥98.0% (by GC)
Tributylsilane
Kaava: C₁₂H₂₈Si
Kiehumispiste: 103°C
Leimahduspiste: 83°C
MDL numero: MFCD00009415
CAS-numero: 998-41-4

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR