Triethylammonium dihydrogentrifluoride ≥95.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

T2022-10G
TCIAT2022-10GEA 46.2 EUR
TCIAT2022-10G
Triethylammonium dihydrogentrifluoride ≥95.0% (by titrimetric analysis)
Triethylammonium dihydrogentrifluoride
Kaava: C₆H₁₈F₃N
Tiheys: 1,094 g/cm³ (20 °C)
MDL numero: MFCD00043294
CAS-numero: 73602-61-6
EINECS: 277-550-5
UN: 2927
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d506fadd5d6d02593da9b4dce18f8fcf55534b4&vid=6d506fadd5d6d02593da9b4dce18f8fcf55534b4&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Red to Green clear liquid
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR