3,4,6-Tri-O-benzyl-2-deoxy-D-glucopyranose ≥98.0% (by HPLC)

Toimittaja: TCI

T1933-100MG
TCIAT1933-100MGEA 80.3 EUR
TCIAT1933-100MG
3,4,6-Tri-O-benzyl-2-deoxy-D-glucopyranose ≥98.0% (by HPLC)
3,4,6-Tri-O-benzyl-2-deoxy-D-glucopyranose
Kaava: C₂₇H₃₀O₅
Storage Temperature: Jääkaappi
MDL numero: MFCD06797170
CAS-numero: 132732-60-6

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 103.0 to 107.0 deg-C
Specific rotation [a]20/D +46.0 to +50.0 deg (C=1, Chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR