Neodymium(III) trifluoromethanesulphonate ≥97.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

T1919-25G T1919-5G
TCIAT1919-25GEA 156 EUR
TCIAT1919-25G TCIAT1919-5G
Neodymium(III) trifluoromethanesulphonate ≥97.0% (by titrimetric analysis)
Neodymium(III) trifluoromethanesulphonate
Kaava: C₃F₉NdO₉S₃
MDL numero: MFCD00192343
CAS-numero: 34622-08-7
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb30c8ab1901404081eaf8bb960c97dc6dfc3d1b&vid=cb30c8ab1901404081eaf8bb960c97dc6dfc3d1b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Amber to Dark purple powder to crystal
Purity (Chelometric Titration) min. 97.0 % (calcd.on anh.substance)
Water max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR