Tribenzylsilane ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

T1916-25G T1916-5G
TCIAT1916-25GEA 296 EUR
TCIAT1916-25G TCIAT1916-5G
Tribenzylsilane ≥98.0% (by GC)
Tribenzylsilane
Kaava: C₂₁H₂₂Si
MDL numero: MFCD00004769
CAS-numero: 1747-92-8

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 88.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR