3-(Trifluorimetyyli)fenyyliboronihappo (contains varying amounts of Anhydride)

Toimittaja: TCI
Varoitus

T1793-1G T1793-5G
TCIAT1793-1GEA 22 EUR
TCIAT1793-1G TCIAT1793-5G
3-(Trifluorimetyyli)fenyyliboronihappo (contains varying amounts of Anhydride)
3-(Trifluorimetyyli)fenyyliboronihappo
Kaava: C₇H₆BF₃O₂
MDL numero: MFCD00151854
CAS-numero: 1423-26-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7b98a61752eb7c8c0582c53e6e51d5324895bbe&vid=f7b98a61752eb7c8c0582c53e6e51d5324895bbe&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR