Scandium(III) trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

T1663-1G T1663-5G
TCIAT1663-1GEA 52.8 EUR
TCIAT1663-1G TCIAT1663-5G
Scandium(III) trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Scandium(III) trifluoromethanesulphonate
Kaava: C₃F₉O₉S₃Sc
MDL numero: MFCD00192433
CAS-numero: 144026-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c321a0d538d42c5ec08e6f73ad98aeeae0d81647&vid=c321a0d538d42c5ec08e6f73ad98aeeae0d81647&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR