Trimethylsilylmethylmagnesium chloride 20% ∼1 mol/L in ethyl ether

Toimittaja: TCI
Vaara

T1451-100ML
TCIAT1451-100MLEA 112 EUR
TCIAT1451-100ML
Trimethylsilylmethylmagnesium chloride 20% ∼1 mol/L in ethyl ether
Trimethylsilylmethylmagnesium chloride
Kaava: C₄H₁₁ClMgSi
MDL numero: MFCD00009916
CAS-numero: 13170-43-9
UN: 3399
ADR: 4.3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=087fa94279748aed40a0a246fafc3fd6e7be4a81&vid=af3d0c3259f580b79f03dab6e165aee006e4c1d8&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Concentration (Neutralization titration) 20.0 to 22.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR