Ethyl-4,4,4-trichloroacetoacetate ≥97.0% (by GC)

Toimittaja: TCI
Varoitus

T1285-10G T1285-25G
TCIAT1285-10GEA 147 EUR
TCIAT1285-10G TCIAT1285-25G
Ethyl-4,4,4-trichloroacetoacetate ≥97.0% (by GC)
Ethyl-4,4,4-trichloroacetoacetate
Kaava: C₆H₇Cl₃O₃
Kiehumispiste: 234°C
MDL numero: MFCD00043329
CAS-numero: 3702-98-5
EINECS: 223-041-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ca058aee52401379c3a0b935cbb2c6239811443&vid=1ca058aee52401379c3a0b935cbb2c6239811443&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR