Ethyl m-tolylacetate ≥98.0% (by GC)

Toimittaja: TCI

T1226-10G
TCIAT1226-10GEA 157 EUR
TCIAT1226-10G
Ethyl m-tolylacetate ≥98.0% (by GC)
Ethyl m-tolylacetate
Kaava: (C₆H₄(CH₃))CH₂COOCH₂CH₃
Kiehumispiste: 238°C
Tiheys: 1,018 g/cm³ (17.5 °C)
MDL numero: MFCD00017273
CAS-numero: 40061-55-0
EINECS: 254-778-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=609ff51f6cbaba83c86239cf809107aff282cb84&vid=609ff51f6cbaba83c86239cf809107aff282cb84&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR