Tetrabutylammonium dichlorobromide ≥97.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

T1186-25G
TCIAT1186-25GEA 344 EUR
TCIAT1186-25G
Tetrabutylammonium dichlorobromide ≥97.0% (by titrimetric analysis)
Tetrabutylammonium dichlorobromide
Kaava: C₁₆H₃₆BrCl₂N
MDL numero: MFCD00060054
CAS-numero: 13053-75-3

Order Now

Testitulosten erittely

Appearance Light yellow to Yellow to Green powder to crystal
Purity (Iodometric Titration) min. 97.0 %
Melting point 63.0 to 67.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR