Methyl-3,4,5-trimethoxybenzoate ≥99.0% (by GC)

Toimittaja: TCI

T1092-25G T1092-500G
TCIAT1092-25GEA 17.6 EUR
TCIAT1092-25G TCIAT1092-500G
Methyl-3,4,5-trimethoxybenzoate ≥99.0% (by GC)
Methyl-3,4,5-trimethoxybenzoate
Kaava: C₁₁H₁₄O₅
Kiehumispiste: 185°C
MDL numero: MFCD00008431
CAS-numero: 1916-07-0
EINECS: 217-629-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57fe3823ead719249f8daeae798b5a5d026fd5c4&vid=57fe3823ead719249f8daeae798b5a5d026fd5c4&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 82.0 to 85.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR