α,α,α-Trifluori-m-tolueenihappo ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

T1034-25G T1034-5G
TCIAT1034-25GEA 88 EUR
TCIAT1034-25G TCIAT1034-5G
α,α,α-Trifluori-m-tolueenihappo ≥98.0% (by GC, titration analysis)
α,α,α-Trifluori-m-tolueenihappo
Kaava: C₈H₅F₃O₂
Kiehumispiste: 126°C
Sulamispiste: 106 °C
Tiheys: 1,371 g/cm³ (25 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002519
CAS-numero: 454-92-2
EINECS: 207-230-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=390fb9714e66580b4c059cf750da4ce2cece8edf&vid=390fb9714e66580b4c059cf750da4ce2cece8edf&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency
Melting point 104.0 to 107.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR