Tris(triphenylphosphine)rhodium(I) chloride ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

T0931-1G
TCIAT0931-1GEA 143 EUR
TCIAT0931-1G
Tris(triphenylphosphine)rhodium(I) chloride ≥98.0% (by titrimetric analysis)
Tris(triphenylphosphine)rhodium(I) chloride
Kaava: C₅₄H₄₅ClP₃Rh
Sulamispiste: 157 °C
Tiheys: 1,363 g/cm³ (20 °C)
MDL numero: MFCD00010016
CAS-numero: 14694-95-2
EINECS: 238-744-5
Merck Index: 14,09758
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88d0c0792907b12f94dab6207ede25bc32f61bbc&vid=88d0c0792907b12f94dab6207ede25bc32f61bbc&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Orange to Brown to Dark red powder to crystal
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR