Tris(4-metyylifenyyli)fosfiini ≥96.0% (by GC)

Toimittaja: TCI
Varoitus

T0862-25G T0862-5G
TCIAT0862-25GEA 124 EUR
TCIAT0862-25G TCIAT0862-5G
Tris(4-metyylifenyyli)fosfiini ≥96.0% (by GC)
Tris(4-metyylifenyyli)fosfiini
Kaava: C₂₁H₂₁P
Kiehumispiste: 400 °C (1013 hPa)
Sulamispiste: 144 °C
Tiheys: 1,11 g/cm³ (20 °C)
MDL numero: MFCD00008542
CAS-numero: 1038-95-5
EINECS: 213-863-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f7f7cc60dad25dfe34104ade91e6a12a54e71fd&vid=9f7f7cc60dad25dfe34104ade91e6a12a54e71fd&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 147.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR