Bis(2,4-pentanedionato)tin(IV) dichloride ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

T0650-25G
TCIAT0650-25GEA 365 EUR
TCIAT0650-25G
Bis(2,4-pentanedionato)tin(IV) dichloride ≥98.0% (by titrimetric analysis)
Bis(2,4-pentanedionato)tin(IV) dichloride
Kaava: C₁₀H₁₄Cl₂O₄Sn
MDL numero: MFCD00015316
CAS-numero: 16919-46-3
EINECS: 240-972-5
UN: 3146
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 204.0 to 208.0 deg-C
Solubility in hot EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR